4-Amino-2-mercaptopyrimidine
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-2-mercaptopyrimidine, 4-Aminopyrimidine-2-thiol, 4-Amino-2-sulphanylpyrimidine
OR1193T-1G OR1193T-250MG
APOSOR1193T-1GEA 87 EUR
APOSOR1193T-1G APOSOR1193T-250MG
4-Amino-2-mercaptopyrimidine
4-Amino-2-mercaptopyrimidine
Formula: C₄H₅N₃S
MW: 127.1676 g/mol
Melting Pt: 273…277 °C
Storage Temperature: Ambient
MDL Number: MFCD00006075
CAS Number: 139424-17-2