2-(2-Methyl-1H-imidazol-1-yl)benzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2-Carboxyphenyl)-2-methyl-1H-imidazole
OR12217-1G OR12217-250MG
APOSOR12217-1GEA 154 EUR
APOSOR12217-1G APOSOR12217-250MG
2-(2-Methyl-1H-imidazol-1-yl)benzoic acid
2-(2-Methyl-1H-imidazol-1-yl)benzoic acid
Formula: C₁₁H₁₀N₂O₂
Melting Pt: 232…234 °C
Storage Temperature: Ambient
MDL Number: MFCD09065012
CAS Number: 159589-71-6
UN: 2811
ADR: 6.1,III