4-(Aminomethyl)-1-methyl-1H-imidazole 97%
Supplier: Apollo Scientific
Danger
Synonyms: (1-Methyl-1H-imidazol-4-yl)methylamine
OR12312-1G OR12312-250MG OR12312-5G
APOSOR12312-1GEA 462 EUR
APOSOR12312-1G APOSOR12312-250MG APOSOR12312-5G
4-(Aminomethyl)-1-methyl-1H-imidazole 97%
4-(Aminomethyl)-1-methyl-1H-imidazole
Formula: C₅H₉N₃
MW: 111.145 g/mol
Boiling Pt: 104…108 °C (0.2 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06411536
CAS Number: 486414-83-9
UN: 2735
ADR: 8,III