4-[4-(Aminomethyl)piperidin-1-yl]thieno[3,2-d]pyrimidine 97%
Supplier: Apollo Scientific
Danger
Synonyms: (1-Thieno[3,2-d]pyrimidin-4-ylpiperid-4-yl)methylamine
OR12375-1G OR12375-250MG
APOSOR12375-1GEA 406 EUR
APOSOR12375-1G APOSOR12375-250MG
4-[4-(Aminomethyl)piperidin-1-yl]thieno[3,2-d]pyrimidine 97%
4-[4-(Aminomethyl)piperidin-1-yl]thieno[3,2-d]pyrimidine
Formula: C₁₂H₁₆N₄S
MW: 248.347 g/mol
Melting Pt: 63.5…72 °C
Storage Temperature: Ambient
MDL Number: MFCD09787494
CAS Number: 937795-94-3
UN: 3259
ADR: 8,III