3-(1-Methyl-1H-pyrazol-5-yl)benzoic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-(3-Carboxyphenyl)-1-methyl-1H-pyrazole
OR12385-1G OR12385-250MG
APOSOR12385-1GEA 372 EUR
APOSOR12385-1G APOSOR12385-250MG
3-(1-Methyl-1H-pyrazol-5-yl)benzoic acid 97%
3-(1-Methyl-1H-pyrazol-5-yl)benzoic acid
Formula: C₁₁H₁₀N₂O₂
MW: 202.209 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD09702398
CAS Number: 628297-55-2
UN: 2811
ADR: 6.1,III