Methyl trans-4-phenylpyrrolidine-3-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: trans-3-(Methoxycarbonyl)-4-phenylpyrrolidine, [trans-4-(Methoxycarbonyl)pyrrolidin-3-yl]benzene

OR12541-1G OR12541-250MG OR12541-5G
APOSOR12541-1GEA 288 EUR
APOSOR12541-1G APOSOR12541-250MG APOSOR12541-5G
Methyl trans-4-phenylpyrrolidine-3-carboxylate
Methyl trans-4-phenylpyrrolidine-3-carboxylate
Formula: C₁₂H₁₅NO₂
MW: 205.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862546
CAS Number: 156469-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR