[3-(4-Chlorophenoxy)phenyl]methylamine hydrochloride 95%
Supplier: Apollo Scientific
Warning
Synonyms: (3-(4-Chlorophenoxy)phenyl)methanamine hydrochloride, 3-(Aminomethyl)-4-chlorodiphenyl ether hydrochloride, 3-(4-Chlorophenoxy)benzylamine hydrochloride
OR12557-250MG OR12557-1G
APOSOR12557-250MGEA 82.8 EUR
APOSOR12557-250MG APOSOR12557-1G
[3-(4-Chlorophenoxy)phenyl]methylamine hydrochloride 95%
[3-(4-Chlorophenoxy)phenyl]methylamine hydrochloride
Formula: C₁₃H₁₂ClNO·ClH
Storage Temperature: Ambient
MDL Number: MFCD02089409
CAS Number: 1171023-50-9
UN: 2811
ADR: 6.1,III