3-Phenoxybenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Chlorosulphonyl)diphenyl ether
OR12558-100MG OR12558-1G OR12558-250MG
APOSOR12558-100MGEA 98.6 EUR
APOSOR12558-100MG APOSOR12558-1G APOSOR12558-250MG
3-Phenoxybenzenesulphonyl chloride
3-Phenoxybenzenesulphonyl chloride
Formula: C₁₂H₉ClO₃S
MW: 268.716 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02089483
CAS Number: 252873-46-4
UN: 3265
ADR: 8 III