3-Acetyl-1-methyl-1H-pyrazole 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(1-Methyl-1H-pyrazol-3-yl)ethan-1-one, 3-Ethanoyl-1-methyl-1H-pyrazole
OR12628-1G OR12628-250MG
APOSOR12628-1GEA 464 EUR
APOSOR12628-1G APOSOR12628-250MG
3-Acetyl-1-methyl-1H-pyrazole 95%
3-Acetyl-1-methyl-1H-pyrazole
Formula: C₆H₈N₂O
MW: 124.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04968783
CAS Number: 137890-04-1