4-(2-Methoxycarbonylethyl)benzeneboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-(2-Methoxycarbonylethyl)phenylboronic acid, (4-(3-Methoxy-3-oxopropyl)phenyl)boronic acid
OR13106-1G OR13106-250MG
APOSOR13106-1GEA 201 EUR
APOSOR13106-1G APOSOR13106-250MG
4-(2-Methoxycarbonylethyl)benzeneboronic acid
4-(2-Methoxycarbonylethyl)benzeneboronic acid
Formula: C₁₀H₁₃BO₄
MW: 208.019 g/mol
Melting Pt: 90…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115640
CAS Number: 850568-44-4