2-Bromo-1,5-dimethyl-1H-imidazole 97%
Supplier: Apollo Scientific
|
Warning
|
OR1343-1G
OR1343-5G
OR1343-100MG
APOSOR1343-1GEA
212
EUR
APOSOR1343-1G
APOSOR1343-5G
APOSOR1343-100MG
2-Bromo-1,5-dimethyl-1H-imidazole 97%
2-Bromo-1,5-dimethyl-1H-imidazole
Formula:
C₅H₇BrN₂ MW: 175.026 g/mol Melting Pt: 40…42 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659902 CAS Number: 235426-31-0 |