[5-(Benzyloxy)-1H-indol-3-yl]acetonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 5-(Benzyloxy)-3-(cyanomethyl)-1H-indole, 5-(Phenylmethoxy)-3-(cyanomethyl)-1H-indole
OR13727-1G OR13727-25G OR13727-5G
APOSOR13727-1GEA 34.8 EUR
APOSOR13727-1G APOSOR13727-25G APOSOR13727-5G
[5-(Benzyloxy)-1H-indol-3-yl]acetonitrile
[5-(Benzyloxy)-1H-indol-3-yl]acetonitrile
Formula: C₁₇H₁₄N₂O
MW: 262.306 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022730
CAS Number: 2436-15-9