2-(4-Bromophenyl)-5-thio-1,3,4-oxadiazole
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Bromophenyl)-5-sulphanyl-1,3,4-oxadiazole, 5-(4-Bromophenyl)-1,3,4-oxadiazole-2-thiol
OR13745-1G OR13745-25G OR13745-5G
APOSOR13745-1GEA 72.5 EUR
APOSOR13745-1G APOSOR13745-25G APOSOR13745-5G
2-(4-Bromophenyl)-5-thio-1,3,4-oxadiazole
2-(4-Bromophenyl)-5-thio-1,3,4-oxadiazole
Formula: C₈H₅BrN₂OS
MW: 257.107 g/mol
Melting Pt: 190…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00992518
CAS Number: 41421-19-6