5-Methylbenzo[b]thiophene-2-boronic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Borono-5-methylbenzo[b]thiophene
OR14304-250MG OR14304-100MG
APOSOR14304-250MGEA 65.4 EUR
APOSOR14304-250MG APOSOR14304-100MG
5-Methylbenzo[b]thiophene-2-boronic acid 97%
5-Methylbenzo[b]thiophene-2-boronic acid
Formula: C₉H₉BO₂S
MW: 192.043 g/mol
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD06200866
CAS Number: 136099-65-5