3-[4-(Hydroxymethyl)phenyl]pyridine 97%
Supplier: Apollo Scientific
Warning
Synonyms: (4-Pyridin-3-ylphenyl)methanol, (4-(Pyridin-3-yl)phenyl)methanol
OR1436-1G OR1436-250MG OR1436-5G
APOSOR1436-1GEA 183 EUR
APOSOR1436-1G APOSOR1436-250MG APOSOR1436-5G
3-[4-(Hydroxymethyl)phenyl]pyridine 97%
3-[4-(Hydroxymethyl)phenyl]pyridine
Formula: C₁₂H₁₁NO
MW: 185.222 g/mol
Melting Pt: 106…107 °C
Storage Temperature: Ambient
MDL Number: MFCD04116293
CAS Number: 217189-04-3