Methyl 3,5-dinitro-2-hydroxybenzoate
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Dinitro-6-(methoxycarbonyl)phenol, Methyl 3,5-dinitrosalicylate
OR14438-1G OR14438-5G
APOSOR14438-1GEA 40.6 EUR
APOSOR14438-1G APOSOR14438-5G
Methyl 3,5-dinitro-2-hydroxybenzoate
Methyl 3,5-dinitro-2-hydroxybenzoate
Formula: C₈H₆N₂O₇
MW: 242.142 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00017561
CAS Number: 22633-33-6