Methyl 4-[(Z)-amino(hydroxyimino)methyl]benzoate 95%
Supplier: Apollo Scientific
Warning
Synonyms: MEthyl 4-[(z)-amino(hydroxyimino)methyl]benzoate
OR1463-1G OR1463-100MG
APOSOR1463-1GEA 30.6 EUR
APOSOR1463-1G APOSOR1463-100MG
Methyl 4-[(Z)-amino(hydroxyimino)methyl]benzoate 95%
Methyl 4-[(Z)-amino(hydroxyimino)methyl]benzoate
Formula: C₉H₁₀N₂O₃
MW: 194.187 g/mol
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD06797503
CAS Number: 184778-33-4