2-Nitro-L-phenylalanine

Supplier: Apollo Scientific
Warning

Synonyms: (2S)-2-Amino-3-(2-nitrophenyl)propionic acid, (2S)-2-Amino-3-(2-nitrophenyl)propanoic acid, (S)-2-Amino-3-(2-nitrophenyl)propanoic acid, H-Phe(2-NO₂)-OH

OR14705-250MG
APOSOR14705-250MGEA 21.9 EUR
APOSOR14705-250MG
2-Nitro-L-phenylalanine
2-Nitro-L-phenylalanine
Formula: C₉H₁₀N₂O₄
MW: 210.187 g/mol
Melting Pt: 226…227 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270360
CAS Number: 19883-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR