6-[(Tetrahydro-2H-pyran-4-yl)oxy]nicotinaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 6-[(Tetrahydro-2H-pyran-4-yl)oxy]pyridine-3-carboxaldehyde, 5-Formyl-2-[(tetrahydro-2H-pyran-4-yl)oxy]pyridine
OR14887-1G OR14887-250MG OR14887-50MG
APOSOR14887-1GEA 169 EUR
APOSOR14887-1G APOSOR14887-250MG APOSOR14887-50MG
6-[(Tetrahydro-2H-pyran-4-yl)oxy]nicotinaldehyde 97%
6-[(Tetrahydro-2H-pyran-4-yl)oxy]nicotinaldehyde
Formula: C₁₁H₁₃NO₃
Melting Pt: 66.5…68 °C
Storage Temperature: Ambient
MDL Number: MFCD09817493
CAS Number: 910036-95-2
UN: 2811
ADR: 6.1,III