4-(3,5-Dimethyl-1H-pyrazol-1-yl)-N-methylbenzylamine 95%
Supplier: Apollo Scientific
Danger
Synonyms: 1-[4-(3,5-Dimethyl-1H-pyrazol-1-yl)phenyl]-N-methylmethylamine, 3,5-Dimethyl-1-{4-[(methylamino)methyl]phenyl}-1H-pyrazole
OR14913-1G OR14913-250MG
APOSOR14913-1GEA 311 EUR
APOSOR14913-1G APOSOR14913-250MG
4-(3,5-Dimethyl-1H-pyrazol-1-yl)-N-methylbenzylamine 95%
4-(3,5-Dimethyl-1H-pyrazol-1-yl)-N-methylbenzylamine
Formula: C₁₃H₁₇N₃
MW: 215.294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08572134
CAS Number: 937796-07-1
UN: 2922
ADR: 8+6.1 III