4-(Piperazin-1-yl)thieno[3,2-d]pyrimidine
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Thieno[3,2-d]pyrimidin-4-yl)piperazine
OR15038-1G OR15038-250MG
APOSOR15038-1GEA 109 EUR
APOSOR15038-1G APOSOR15038-250MG
4-(Piperazin-1-yl)thieno[3,2-d]pyrimidine
4-(Piperazin-1-yl)thieno[3,2-d]pyrimidine
Formula: C₁₀H₁₂N₄S
MW: 220.294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407481
CAS Number: 683274-67-1