N-[(4-Cyanophenyl)carbamothioyl]benzamide
Supplier: Apollo Scientific
Warning
Synonyms: 4-[(Benzoylcarbamothioyl)amino]benzonitrile, 1-Benzoyl-3-(4-cyanophenyl)thiourea
OR15041-5G OR15041-10G OR15041-25G
APOSOR15041-5GEA 52.2 EUR
APOSOR15041-5G APOSOR15041-10G APOSOR15041-25G
N-[(4-Cyanophenyl)carbamothioyl]benzamide
N-[(4-Cyanophenyl)carbamothioyl]benzamide
Formula: C₁₅H₁₁N₃OS
MW: 281.332 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00587941
CAS Number: 1448-64-2