5-Chloro-2-(1H-tetrazol-5-yl)aniline 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-(2-Amino-4-chlorophenyl)-1H-tetrazole
OR15044-1G OR15044-25G OR15044-5G
APOSOR15044-1GEA 27.7 EUR
APOSOR15044-1G APOSOR15044-25G APOSOR15044-5G
5-Chloro-2-(1H-tetrazol-5-yl)aniline 97%
5-Chloro-2-(1H-tetrazol-5-yl)aniline
Formula: C₇H₆ClN₅
MW: 195.609 g/mol
Melting Pt: 160…162 °C
Storage Temperature: Ambient
MDL Number: MFCD03085917
CAS Number: 54013-18-2