2-Bromo-4,5-dimethoxycinnamic acid 97%
Supplier: Apollo Scientific
OR1565-25G OR1565-50G
APOSOR1565-25GEA 122 EUR
APOSOR1565-25G APOSOR1565-50G
2-Bromo-4,5-dimethoxycinnamic acid 97%
2-Bromo-4,5-dimethoxycinnamic acid
Formula: C₁₀H₉BrO₃
MW: 287.12 g/mol
Melting Pt: 253…256 °C
Storage Temperature: Ambient
MDL Number: MFCD00017614
CAS Number: 195383-80-3