5-(Bromomethyl)-3-methylisoxazole 95%

Supplier: Apollo Scientific
Danger

Synonyms: 5-(Bromomethyl)-3-methyl-1,2-oxazole

OR15946-250MG OR15946-1G
APOSOR15946-250MGEA 120 EUR
APOSOR15946-250MG APOSOR15946-1G
5-(Bromomethyl)-3-methylisoxazole 95%
5-(Bromomethyl)-3-methylisoxazole
Formula: C₅H₆BrNO
MW: 176.011 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09879901
CAS Number: 36958-61-9
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR