5-Amino-1,3-thiazole
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Thiazol-5-amine
OR16619-250MG OR16619-1G
APOSOR16619-250MGEA 82.8 EUR
APOSOR16619-250MG APOSOR16619-1G
5-Amino-1,3-thiazole
5-Amino-1,3-thiazole
Formula: C₃H₄N₂S
MW: 100.142 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09834069
CAS Number: 17721-00-5