Sodium 2,4-dichlorobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Dichlorobenzoic acid sodium salt
OR16639-100G OR16639-25G OR16639-500G
APOSOR16639-100GEA 132 EUR
APOSOR16639-100G APOSOR16639-25G APOSOR16639-500G
Sodium 2,4-dichlorobenzoate
Sodium 2,4-dichlorobenzoate
Formula: C₇H₃Cl₂O₂·Na
MW: 212.993 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606076
CAS Number: 38402-11-8