1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)-1H-pyrrole
Supplier: Apollo Scientific
Synonyms: 1-(2,3-Dihydro-7-nitro-1,1,3,3,6-pentamethyl-1H-inden-5-yl)-1H-pyrrole
OR16908-1G OR16908-5G
APOSOR16908-1GEA 174 EUR
APOSOR16908-1G APOSOR16908-5G
1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)-1H-pyrrole
1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)-1H-pyrrole
Formula: C₁₈H₂₂N₂O₂
MW: 298.379 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00206198
CAS Number: 657405-94-2