1H,2H,2H-Perchloropropane
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,1,3,3-Pentachloropropane, 240fa
OR16934-25G
APOSOR16934-25GEA 212 EUR
APOSOR16934-25G
1H,2H,2H-Perchloropropane
1H,2H,2H-Perchloropropane
Formula: C₃H₃Cl₅
MW: 216.321 g/mol
Boiling Pt: 74 °C ( 28 mm Hg)
Density: 1.597 g/cm³ (25 °C)
Flash Pt: 69.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00045255
CAS Number: 23153-23-3