4-(Hydroxymethyl)-1H-benzimidazole 97%
Supplier: Apollo Scientific
Warning
Synonyms: (1H-Benzimidazol-4-yl)methanol, 4-(Hydroxymethyl)benzimidazole
OR17072-250MG OR17072-1G
APOSOR17072-250MGEA 168 EUR
APOSOR17072-250MG APOSOR17072-1G
4-(Hydroxymethyl)-1H-benzimidazole 97%
4-(Hydroxymethyl)-1H-benzimidazole
Formula: C₈H₈N₂O
MW: 148.162 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15144561
CAS Number: 65658-13-1