4-(Bromomethyl)-5-methyl-1,3-dioxol-2-one
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Bromomethyl)-5-methyl-2-oxo-1,3-dioxole, 4-Bromomethyl-5-methyl-1,3-dioxol-2-one
OR17618-1G OR17618-250MG
APOSOR17618-1GEA 403 EUR
APOSOR17618-1G APOSOR17618-250MG
4-(Bromomethyl)-5-methyl-1,3-dioxol-2-one
4-(Bromomethyl)-5-methyl-1,3-dioxol-2-one
Formula: C₅H₅BrO₃
MW: 192.995 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07787463
CAS Number: 80715-22-6