2-Amino-4-iodo-1,3-benzothiazole
Supplier: Apollo Scientific
Warning
Synonyms: 4-Iodo-1,3-benzothiazol-2-amine
OR17932-1G OR17932-250MG
APOSOR17932-1GEA 763 EUR
APOSOR17932-1G APOSOR17932-250MG
2-Amino-4-iodo-1,3-benzothiazole
2-Amino-4-iodo-1,3-benzothiazole
Formula: C₇H₅IN₂S
MW: 276.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11194508
CAS Number: 1039326-79-8