2-Chloro-4-methylpyrimidine-5-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-4-methyl-1,3-diazine-5-carboxylic acid, 5-Carboxy-2-chloro-4-methylpyrimidine
OR18025-1G OR18025-250MG
APOSOR18025-1GEA 304 EUR
APOSOR18025-1G APOSOR18025-250MG
2-Chloro-4-methylpyrimidine-5-carboxylic acid
2-Chloro-4-methylpyrimidine-5-carboxylic acid
Formula: C₆H₅ClN₂O₂
MW: 172.569 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09863167
CAS Number: 188781-10-4