6-Chloro-2-methyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Supplier: Apollo Scientific
OR183446-1G OR183446-250MG
APOSOR183446-1GEA 364 EUR
APOSOR183446-1G APOSOR183446-250MG
6-Chloro-2-methyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
6-Chloro-2-methyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Formula: C₁₁H₉ClN₄
MW: 232.669 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269630
CAS Number: 372198-32-8