4-(Morpholin-4-yl)benzene-1,2-diamine
Supplier: Apollo Scientific
OR183524-1G OR183524-250MG
APOSOR183524-1GEA 551 EUR
APOSOR183524-1G APOSOR183524-250MG
4-(Morpholin-4-yl)benzene-1,2-diamine
4-(Morpholin-4-yl)benzene-1,2-diamine
Formula: C₁₀H₁₅N₃O
MW: 193.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848955
CAS Number: 119421-28-2