Methyl 2-hydroxy-4-nitrobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4-nitrosalicylate, 3-Hydroxy-4-(methoxycarbonyl)nitrobenzene
OR18576-1G OR18576-250MG OR18576-5G
APOSOR18576-1GEA 136 EUR
APOSOR18576-1G APOSOR18576-250MG APOSOR18576-5G
Methyl 2-hydroxy-4-nitrobenzoate
Methyl 2-hydroxy-4-nitrobenzoate
Formula: C₈H₇NO₅
MW: 197.145 g/mol
Melting Pt: 101…102 °C
Storage Temperature: Ambient
MDL Number: MFCD11041295
CAS Number: 13684-28-1