2,6-Dichloro-4-methylaniline
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dichloro-p-toluidine, 4-Amino-3,5-dichlorotoluene, 2,6-Dichloro-4-toluidine
OR19488-1G OR19488-5G
APOSOR19488-1GEA 68.3 EUR
APOSOR19488-1G APOSOR19488-5G
2,6-Dichloro-4-methylaniline
2,6-Dichloro-4-methylaniline
Formula: C₇H₇Cl₂N
MW: 176.043 g/mol
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00190134
CAS Number: 56461-98-4