4-(4-Bromophenyl)butan-2-ol
Supplier: Apollo Scientific
OR200157-1G OR200157-250MG OR200157-500MG OR200157-2.5G
APOSOR200157-1GEA 519 EUR
APOSOR200157-1G APOSOR200157-250MG APOSOR200157-500MG APOSOR200157-2.5G
4-(4-Bromophenyl)butan-2-ol
4-(4-Bromophenyl)butan-2-ol
Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16150637
CAS Number: 223554-16-3