[2-(Pyrrolidin-1-yl)pyridin-4-yl]methanol 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 1-[4-(Hydroxymethyl)pyridin-2-yl]pyrrolidine, 4-(Hydroxymethyl)-2-(pyrrolidin-1-yl)pyridine
OR2058-1G OR2058-5G
APOSOR2058-1GEA 487 EUR
APOSOR2058-1G APOSOR2058-5G
[2-(Pyrrolidin-1-yl)pyridin-4-yl]methanol 97+%
[2-(Pyrrolidin-1-yl)pyridin-4-yl]methanol
Formula: C₁₀H₁₄N₂O
MW: 178.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741433
CAS Number: 906352-65-6
UN: 2811
ADR: 6.1,III