2,4-Bis(tert-butyl)-6-nitrophenol
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Bis(tert-butyl)-2-hydroxynitrobenzene
OR21278-1G OR21278-250MG OR21278-5G
APOSOR21278-1GEA 184 EUR
APOSOR21278-1G APOSOR21278-250MG APOSOR21278-5G
2,4-Bis(tert-butyl)-6-nitrophenol
2,4-Bis(tert-butyl)-6-nitrophenol
Formula: C₁₄H₂₁NO₃
MW: 251.321 g/mol
Melting Pt: 59…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00026282
CAS Number: 20039-94-5