2-[(4-Chlorophenyl)thio]-5-nitrobenzonitrile
Supplier: Apollo Scientific
OR21637-1G OR21637-5G
APOSOR21637-1GEA 87 EUR
APOSOR21637-1G APOSOR21637-5G
2-[(4-Chlorophenyl)thio]-5-nitrobenzonitrile
2-[(4-Chlorophenyl)thio]-5-nitrobenzonitrile
Formula: C₁₃H₇ClN₂O₂S
MW: 290.725 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067807
CAS Number: 78940-73-5