Ethyl 3-(2-chloro-3,4-dimethoxyphenyl)acrylate
Supplier: Apollo Scientific
OR21780-1G OR21780-5G
APOSOR21780-1GEA 79.9 EUR
APOSOR21780-1G APOSOR21780-5G
Ethyl 3-(2-chloro-3,4-dimethoxyphenyl)acrylate
Ethyl 3-(2-chloro-3,4-dimethoxyphenyl)acrylate
Formula: C₁₃H₁₅ClO₄
MW: 270.709 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085063
CAS Number: 175135-96-3