1-Isobutyl-1H-benzimidazole
Supplier: Apollo Scientific
Warning
OR2267-1G
APOSOR2267-1GEA 55.1 EUR
APOSOR2267-1G
1-Isobutyl-1H-benzimidazole
1-Isobutyl-1H-benzimidazole
Formula: C₁₁H₁₄N₂
Boiling Pt: 155…165 °C
Melting Pt: 49…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00464042
CAS Number: 305346-88-7