3-(Methoxycarbonyl)-5-[methyl(methylsulphonyl)amino]benzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methyl 5-[methyl(methylsulphonyl)amino]isophthalate, N-[3-Carboxy-5-(methoxycarbonyl)phenyl]-N-methylmethanesulphonamide
OR2286-100MG OR2286-500MG
APOSOR2286-100MGEA 116 EUR
APOSOR2286-100MG APOSOR2286-500MG
3-(Methoxycarbonyl)-5-[methyl(methylsulphonyl)amino]benzoic acid
3-(Methoxycarbonyl)-5-[methyl(methylsulphonyl)amino]benzoic acid
Formula: C₁₁H₁₃NO₆S
MW: 287.289 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954475
CAS Number: 695215-94-2