4-[(4-Methoxyphenyl)amino]piperidine, N1-BOC protected
Supplier: Apollo Scientific
Synonyms: tert-Butyl 4-(4-methoxyanilino)tetrahydropyridine-1-carboxylate
OR22955-1G OR22955-5G
APOSOR22955-1GEA 87 EUR
APOSOR22955-1G APOSOR22955-5G
4-[(4-Methoxyphenyl)amino]piperidine, N1-BOC protected
4-[(4-Methoxyphenyl)amino]piperidine, N1-BOC protected
Formula: C₁₇H₂₆N₂O₃
MW: 306.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02102341
CAS Number: 306934-84-9