(5-Methyl-1-phenyl-1H-pyrazol-4-yl)methanol
Supplier: Apollo Scientific
Warning
OR23248-1G OR23248-250MG
APOSOR23248-1GEA 88.6 EUR
APOSOR23248-1G APOSOR23248-250MG
(5-Methyl-1-phenyl-1H-pyrazol-4-yl)methanol
(5-Methyl-1-phenyl-1H-pyrazol-4-yl)methanol
Formula: C₁₁H₁₂N₂O
Melting Pt: 88…89 °C
Storage Temperature: Ambient
MDL Number: MFCD02682046
CAS Number: 153863-35-5