5-Methyl-1-phenyl-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR23251-1G OR23251-5G
APOSOR23251-1GEA 130 EUR
APOSOR23251-1G APOSOR23251-5G
5-Methyl-1-phenyl-1H-pyrazole
5-Methyl-1-phenyl-1H-pyrazole
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 70…80 °C (0.1 mm Hg)
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD03086113
CAS Number: 6831-91-0