6-(Morpholin-4-yl)nicotinic acid hydrazide
Supplier: Apollo Scientific
Warning
Synonyms: 6-(4-Morpholino)nicotinohydrazide
OR23277-1G OR23277-250MG
APOSOR23277-1GEA 696 EUR
APOSOR23277-1G APOSOR23277-250MG
6-(Morpholin-4-yl)nicotinic acid hydrazide
6-(Morpholin-4-yl)nicotinic acid hydrazide
Formula: C₁₀H₁₄N₄O₂
MW: 222.244 g/mol
Melting Pt: 196…198 °C
Storage Temperature: Ambient
MDL Number: MFCD03407297
CAS Number: 388088-71-9