4-(1H-Pyrazol-1-yl)benzoyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 1-[4-(Chlorocarbonyl)phenyl]-1H-pyrazole
OR23289-1G OR23289-250MG
APOSOR23289-1GEA 221 EUR
APOSOR23289-1G APOSOR23289-250MG
4-(1H-Pyrazol-1-yl)benzoyl chloride
4-(1H-Pyrazol-1-yl)benzoyl chloride
Formula: C₁₀H₇ClN₂O
MW: 206.628 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02681936
CAS Number: 220461-83-6
UN: 3261
ADR: 8,III