6,7-Dimethoxyisochroman-3-one
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Dihydro-6,7-dimethoxy-3-oxo-1H-isochromene, 3,4-Dihydro-6,7-dimethoxy-3-oxo-1H-2-benzopyran
OR23500-1G OR23500-5G
APOSOR23500-1GEA 140 EUR
APOSOR23500-1G APOSOR23500-5G
6,7-Dimethoxyisochroman-3-one
6,7-Dimethoxyisochroman-3-one
Formula: C₁₁H₁₂O₄
MW: 208.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011571
CAS Number: 16135-41-4